Would you like to log in again?
Human Recombinant PRDX 3, BioVision
Supplier: BioVision
6320-100
10008-178
Human Recombinant PRDX 3, BioVision
>95% Pure Recombinant Active Human PRDX 3
Specific activity: approximately 82-83 pmole/min/µg.
Peroxiredoxin-3 (Prdx3) is a mitochondrial member of the antioxidant family of thioredoxin peroxidases that uses mitochondrial thioredoxin-2 (Trx2) as a source of reducing equivalents to scavenge hydrogen peroxide (H2O2). It acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty