Would you like to log in again?
Human Recombinant UCHL3, BioVision
Supplier: BioVision
6358-100
10008-408EA 334.75 USD InStock
10008-408
Human Recombinant UCHL3, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human UCHL3.
Specific activity: >3000 pmole/min/µg. Measured by the hydrolysis of Ubiquitin-AMC at pH 8.0, at 37°C.
Ubiquitin C-terminal hydrolases (UCHs) are a family of cysteine hydrolases that catalyze the hydrolysis of amides, esters and thioesters of the C-terminus of ubiquitin. UCH-L3 is a member of the lower molecular weight group of UCHs involved in the hydrolysis of small C-terminal derivatives of ubiquitin that form non-specifically during the process of protein ubiquitinylation. UCHL3 play a role in the regulation of neuronal development and spermatogenesis and have been implicated in neurodegenerative diseases.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty