Would you like to log in again?
Human Recombinant MDH2, BioVision
Supplier: BioVision
6356-50
10008-404EA 420.01 USD InStock
10008-404
Human Recombinant MDH2, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human MDH2.
Specific activity is > 30 units/mg, and is defined as the amount of enzyme that cleaves 1 µmole of oxaloacetate and beta-NADH to L-malate and beta-NAD per minute at pH 7.5 at 25°C.
MDH2 catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. MDH2 is localized to the mitochondria and takes part in the malate-aspartate shuttle that functions in the metabolic coordination between cytosol and mitochondria. MDH2 is highly expressed in the adrenal system, small intestine, heart and pancreas.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty