Would you like to log in again?
Human Recombinant FKBP14, BioVision
Supplier: BioVision
6354-100
10008-400EA 355.35 USD InStock
10008-400
Human Recombinant FKBP14, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human FKBP14.
Specific activity is > 240 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1 µmole of Suc-AAFP-pNA per minute at 25°C in Tris HCl pH 8.0 using chymotrypsin.
FKBP14 enzyme accelerates the folding of proteins during protein synthesis. It contains 2 EF-hand domains and one PPIase FKBP-type domain. Truncation of the amino-terminus of FKBP14 significantly decreases peptidyl prolyl cis-trans isomerase activity, therefore suggesting that the PPIase FKBP-type domain must be located at the N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty