Would you like to log in again?
Human Recombinant GSR, BioVision
Supplier: BioVision
6352-50
10008-396
Human Recombinant GSR, BioVision
>95% Pure Recombinant Active Human GSR.
Specific activity: > 29 units/ml.
Glutathione reductase, also known as GSR or GR, is an enzyme that reduces glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. In most eukaryotic cells, GSR maintains the ratio of GSH / GSSG, and participates in several
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty