Would you like to log in again?
Schistosoma japonicum Recombinant GST, BioVision
Supplier: BioVision
6351-100
10008-394
Schistosoma japonicum Recombinant GST, BioVision
>95% Pure Recombinant Active Schistosoma japonicum GST.
Specific activity is 2.8 – 3.3 units/mg
Glutathione S-transferase (GST) represents a major group of detoxification enzymes. It acts by catalyzing the reaction of glutathione with an acceptor molecule to form an S-substituted glutathione (S=sulfur). The reactions utilizing glutathione contribute to the transformation of a wide range of compounds, including carcinogens, therapeutic drugs, and products of oxidative stress. Apart from its enzymatic activities, GST may also bind toxins and function as a transport protein.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty