Would you like to log in again?
Human Recombinant Thioredoxin 2, BioVision
Supplier: BioVision
6318-100
10008-174EA 417.15 USD InStock
10008-174
Human Recombinant Thioredoxin 2, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human Thioredoxin 2
Specific activity is 3-4 A650/min/mg.
Thioredoxin 2 also known as mitochondrial Thioredoxin in humans is encoded by the TXN2 gene. This nuclear gene encodes a mitochondrial member of the thioredoxin family, a group of small multifunctional redox-active proteins. The encoded protein may play important roles in the regulation of the mitochondrial membrane potential and in protection against oxidant-induced apoptosis.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty