Mouse Recombinant Gstmu 1 (from E. coli)
Supplier: BioVision
AB168035-100UG
10008-388EA 873.33 USD
10008-388
Mouse Recombinant Gstmu 1 (from E. coli)
Proteins and Peptides
>95% Pure Recombinant Active Murine GSTMu1.
Specific activity is < 11 units/mg.
GSTM1 is a glutathione S-transferase that belongs to the mu class. This enzyme acts by catalyzing the reaction of glutathione with an acceptor molecule to form an S-substituted glutathione (S=sulfur). The reactions utilizing glutathione contribute the transformation of a wide variety of electrophiles, including reactive products of lipid, protein, carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress.