Human Recombinant Gsta1 (from E. coli)
Supplier: BioVision
AB167981-100UG
10008-384EA 864.37 USD
10008-384
Human Recombinant Gsta1 (from E. coli)
Proteins and Peptides
>90% Pure Recombinant Active Human GSTA1.
Specific activity is 5-8 units/mg.
Glutathione S-transferase A1 in humans is encoded by the GSTA1 gene. Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. GSTA1 protects the cells from reactive oxygen species and is highly expressed in liver.