Would you like to log in again?
Human Recombinant Cyclophilin G, BioVision
Supplier: BioVision
6316-100
10008-170
Human Recombinant Cyclophilin G, BioVision
>95% Pure Recombinant Active Human Cyclophilin G
Specific activity is > 200 nmoles/min/mg.
Peptidyl-prolyl cis-trans isomerase G (PPIG), also known as Cyclophilin G, is a member of peptidylprolyl cis-trans isomerase family (PPIases). This protein catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and is implicated in the folding, transport, and assembly of proteins. It is localized to the nuclear speckles, a nuclear compartment rich in splicing factors, and interacts with the splicing factors SC35 and pinin. Cyclophilin G also may play an important role in the regulation of pre-mRNA splicing.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty