Would you like to log in again?
Human Recombinant FKBPL, BioVision
Supplier: BioVision
6345-10
10008-382EA 386.25 USD InStock
10008-382
Human Recombinant FKBPL, BioVision
Proteins and Peptides
>90% Pure Recombinant Active Human FKBPl.
Specific activity is > 210 nmoles/min/mg.
FKBPL is related to the immunophilin protein family, which take part in immunoregulation and regular cellular processes involving protein folding and trafficking. FKBPL has an important function in the induced radio resistance and participates in the control of the cell cycle. FKBPL has a role in cellular response to stress. FKBPL interacts with Hsp90, glucocorticoid receptor and dynamitin and is involved in signaling, like other FKBPs.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty