Would you like to log in again?
Human Recombinant FKBP6, BioVision
Supplier: BioVision
6344-50
10008-380
Human Recombinant FKBP6, BioVision
>95% Pure Recombinant Active Human FKBP6.
Specific activity is > 190 nmoles/min/mg.
FKBP6 is part of the immunophilin protein family, and is involved in immunoregulation and basic cellular functions involving protein folding and trafficking. FKBP6 is widely expressed in all tissues, FKBP6 is present at utmost levels in testis, liver, kidney, skeletal muscle and heart. Deletion of FKBP6 contributes to hypercalcemia and growth delay in Williams-Beuren syndrome.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty