Would you like to log in again?
Human Recombinant Cyclophilin E, BioVision
Supplier: BioVision
6314-100
10008-166EA 417.15 USD InStock
10008-166
Human Recombinant Cyclophilin E, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human Cyclophilin E
Specific activity is > 210 nmoles/min/mg
Cyclophilins possess the activity of peptidyl-prolyl cis-trans isomerase (PPIase) and are involved in cellular protein folding and protein interactions. PPIE contains two RNA binding domains at the N-terminal region and a PPIase domain at the C-terminal region. The cellular and physiological roles of PPIE are still not known.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty