Human Recombinant Cyclophilin H (from E. coli)
Supplier: BioVision
6313-100
10008-164EA 472.83 USD
10008-164
Human Recombinant Cyclophilin H (from E. coli)
>95% Pure Recombinant Active Human Cyclophilin H
Specific activity is > 220 nmoles/min/mg
Cyclophilin H is a member of peptidyl-propyl cis-trans isomerase (PPIase) family, which catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins. The cyclophilin H is a specific component of the human U4/U6 small nuclear ribonucleoprotein particle involved in the nuclear splicing of pre-mRNA. It stably associates with the U4/U6-60kD and -90kD proteins, the human orthologues of the Saccharomyces cerevisiae Prp4 and Prp3 splicing factors.