Would you like to log in again?
Human Recombinant Cyclophilin D, BioVision
Supplier: BioVision
6312-100
10008-162
Human Recombinant Cyclophilin D, BioVision
>95% Pure Recombinant Active Human Cyclophilin D
Specific activity is > 210 nmoles/min/mg
Cyclophilin D is a member of peptidyl-propyl cis-trans isomerase (PPIase) family, which catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins and thus their formation. Cyclophilin D is also a mitochondrial matrix protein, which plays a decisive role in mitochondrial permeability transition. It can bind to the immunosuppressant cyclosporine A and its overexpression suppresses the apoptosis in cancer cells.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty