Would you like to log in again?
Human Recombinant Cyclophilin A, BioVision
Supplier: BioVision
6311-100
10008-160EA 334.75 USD InStock
10008-160
Human Recombinant Cyclophilin A, BioVision
Proteins and Peptides
>95% Pure Recombinant Active Human Cyclophilin A
Specific activity is > 270 nmoles/min/mg
Cyclophilin A is a multifunctional protein that is upregulated in a variety of inflammatory conditions, such as rheumatoid arthritis, autoimmune disease, and cancer. It has been classified as an immunophilin and has a variety of intracellular functions, including intracellular signaling, protein trafficking, and the regulation of other proteins activity. Besides its intracellular functions, Cyclophilin A is a secreted molecule that has a physiological and pathological role in cardiovascular diseases, making it a potential biomarker and mediator in cardiovascular diseases, such as vascular stenosis, atherosclerosis, and abdominal aortic aneurysms.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty