Human Recombinant UCHL1 (from E. coli)
Supplier: BioVision
AB285718-100UG
10008-348EA 988.25 USD
10008-348
Human Recombinant UCHL1 (from E. coli)
Proteins and Peptides
>95% Pure Recombinant Human PTP1B
> 300 pmole/min/ug
PGP9.5/UCH-L1 is a member of a gene family whose products hydrolyze small C-terminal adducts of ubiquitin to generate the ubiquitin monomer. PGP9.5 is a component of the ubiquitin system, which has a fundamental role in regulating various biological activities. PGP9.5 gene encodes two opposing enzymatic activities that affect alpha-synuclein degradation and Parkinson's disease susceptibility. Recombinant PGP9.5 protein was expressed in E.coli and purified by using conventional chromatography techniques.