Would you like to log in again?
Human Recombinant Thioredoxin, BioVision
Supplier: BioVision
6305-100
10008-346EA 284.1 USD InStock
10008-346
Human Recombinant Thioredoxin, BioVision
Proteins and Peptides
>95% Pure Recombinant Human PTP1B
7-10 A650/min/mg
Thioredoxin (Trx) is a low molecular weight redox protein. Trx contains a redox active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. It is involved in the first unique step in DNA synthesis. Trx also provides control over a number of transcription factors affecting cell proliferation and death through a mechanism referred to as redox regulation. Recombinant human thioredoxin-1 was overexpressed in E. coli and purified by using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty