Would you like to log in again?
Human Recombinant PP2C alpha, BioVision
Supplier: BioVision
6303-100
10008-342EA 288.4 USD InStock
10008-342
Human Recombinant PP2C alpha, BioVision
Proteins and Peptides
>95% Pure Recombinant Human PTP1B
>8,000 units/mg.
Protein phosphatase 2C (PP2C alpha) is a Mn2+- or Mg2+- dependent protein serine/threonine phosphatase that is essential for regulating cellular stress response in eukaryotes. The protein coding region of PP2C alpha (amino acids 1-382) was cloned into an E. coli expression vector. PP2C alpha was overexpressed in E. coli as a soluble His-tag fusion protein, and it was purified by conventional column chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty