Would you like to log in again?
Human Recombinant SHP-1, BioVision
Supplier: BioVision
6302-100
10008-340EA 475.33 USD InStock
10008-340
Human Recombinant SHP-1, BioVision
Proteins and Peptides
>95% Pure Recombinant Human SHP-1
>5,000 units/mg of SHP-1.
The protein coding region of the catalytic domain of SHP-1 (amino acids 243-541) was cloned into an E. coli expression vector. The catalytic domain of SHP-1 was overexpressed as insoluble protein aggregates (inclusion bodies). The recombinant SHP-1 protein was purified by FPLC gel-filtration chromatography, after refolding of the isolated inclusion bodies in a redox buffer. Additional amino acid (Met) is attached at N-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty