Would you like to log in again?
NT-4, human recombinant, BioVision
Supplier: BioVision
4309-10 4309-50 4309-1000
10006-516 10006-520 10006-518
NT-4, human recombinant, BioVision
>98% Pure active Recombinant human NT-4, a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells.
The ED₅₀ as determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures was found in the range of 20-50 ng/ml.
Neurotrophin-4 (NT-4/NT-5) is a potent neurotrophic factor that supports the growth and survivability of nerve and/or glial cells. The active form of human NT-4 is a dimer, formed by two identical 130 amino acid subunits.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty