Would you like to log in again?
BTC, human recombinant, BioVision
Supplier: BioVision
4876-20 4876-1000
10007-258EA 242.05 USD InStock
10007-258 10007-256
BTC, human recombinant, BioVision
Proteins and Peptides
>97% Pure active Recombinant human BTC.
The ED₅₀, calculated by the dose-dependant proliferation of murine BALB\C 3T3 cells (measured by 3H-thymidine uptake) is < 0.05 ng/ml; corresponding to a Specific Activity of > 2.0 ×10⁷IU/mg.
Human Betacellulin (BTC) is a potent mitogen for retinal pigment epithelial cells and vascular smooth muscle cells. The effects of betacellulin are probably mediated by the egf receptor and other related receptors. Human Recombinant BTC produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 80 amino acids and having a molecular mass of 9 kDa. BTC was purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty