Would you like to log in again?
PRMT1, mouse recombinant (Active), BioVision
Supplier: BioVision
4868-1000 4868-50 4868-10
10007-248 10007-244 10007-246
PRMT1, mouse recombinant (Active), BioVision
>90% Pure active Recombinant murine PRMT1.
PRMT1 methylate’s (mono & asymmetric dimethylation) the guanidino nitrogens of arginyl residues present in a glycine and arginine-rich domain (may methylate HNRNPA1 and histones) methylate’s SUPT5H. The PRMT1 protein functions as a histone methyltransferase specific for H4. PRMT1 is an essential factor in oncogenesis and is a potential novel therapeutic target in cancer. PRMT1-mediated methylation serves as a positive modulator of IR/IRS-1/PI3K pathway and glucose uptake in skeletal muscle cells. CAF1 is a new regulator of PRMT1-dependent arginine methylation. PRMT1 arginine-methylate’s MRE11 therefore it regulates the activity of MRE11-RAD50-NBS1 complex during the intra-S-phase DNA damage checkpoint response. PRMT1 plays a post-translationally part in regulating the transcriptional activity. PRMT1 is found predominantly in the cytoplasm, though a fraction of PRMT1 is located in the nucleus.
PRMT1 Mouse Recombinant fused with His-MBP tag at N-terminus produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 750 amino acids and having a molecular mass of 84 kDa. The PRMT1 is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty