Human Recombinant PRMT1 (active) (from E. coli)
4865-10 4865-50 4865-1000
10007-238EA 110.91 USD
10007-238 10007-240 10007-242
Human Recombinant PRMT1 (active) (from E. coli)
>90% Pure active Recombinant human PRMT1.
PRMT1 methylate’s (mono & asymmetric dimethylation) the guanidino nitrogens of arginyl residues present in a glycine and arginine-rich domain (may methylate HNRNPA1 and histones) methylate’s SUPT5H. The PRMT1 protein functions as a histone methyltransferase specific for H4. PRMT1 is an essential factor in oncogenesis and is a potential novel therapeutic target in cancer. PRMT1-mediated methylation serves as a positive modulator of IR/IRS-1/PI3K pathway and glucose uptake in skeletal muscle cells. CAF1 is a new regulator of PRMT1-dependent arginine methylation. PRMT1 arginine-methylate’s MRE11 therefore it regulates the activity of MRE11-RAD50-NBS1 complex during the intra-S-phase DNA damage checkpoint response. PRMT1 plays a post-translationally part in regulating the transcriptional activity. PRMT1 is found predominantly in the cytoplasm, though a fraction of PRMT1 is located in the nucleus.
PRMT1 Human Recombinant (a.a. 1-353) fused with His-MBP tag at N-terminus produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 750 amino acids and having a molecular mass of 84 kDa. The PRMT1 is purified by proprietary chromatographic techniques.