Would you like to log in again?
CRP, human recombinant, BioVision
Supplier: BioVision
4863-10 4863-50 4863-1000
10007-228 10007-232 10007-230
CRP, human recombinant, BioVision
>95% Pure Recombinant human CRP.
Recombinant Human C-Reactive Protein produced in E. coli is a non-glycosylated polypeptide chain having a total molecular mass of 115kDa, corresponding to the pentamer structure of 23 kDa monomers determined by amino acid sequence. Recombinant Human CRP is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty