Would you like to log in again?
Isopeptidase T (long form), human recombinant, BioVision
Supplier: BioVision
4862-25
10006-850EA 211.15 USD InStock
10006-850
Isopeptidase T (long form), human recombinant, BioVision
Enzymes
>95% Pure Recombinant human Isopeptidase T (long form).
Isopeptidase T is a ubiquitin-specific processing protease (UBP) that specifically disassembles free K48-linked polyubiquitin chains into ubiquitin monomers. There are two human isopeptidase T isoforms; Isopeptidase T short form (isoT-S) and Isopeptidase T long form (isoT-L) which differ by an insertion of 23 amino acids in isoT-L. Isopeptidase T is the only UBP known to require zinc binding to be active.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty