Would you like to log in again?
Isopeptidase T (short form), human recombinant, BioVision
Supplier: BioVision
4861-25
10006-848
Isopeptidase T (short form), human recombinant, BioVision
>95% Pure Recombinant human Isopeptidase T (short form).
Ubiquitin C-terminal hydrolases (UCHs) are a family of cysteine hydrolases that catalyze the UBPs hydrolyze isopeptide bonds between ubiquitin and the substrate protein or within polyubiquitin chains. Human isopeptidase T (isoT) is an UBP capable of binding ubiquitin that specifically disassembles free polyubiquitin chains linked through isopeptide bonds involving the K48 of ubiquitin into ubiquitin monomers.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty