Would you like to log in again?
IFN-beta 1b, human recombinant, BioVision
Supplier: BioVision
4860-10 4860-50 4860-1000
10006-842 10006-846 10006-844
IFN-beta 1b, human recombinant, BioVision
>98% Pure active Recombinant human IFN-beta 1b.
BioVision’s Interferon β 1b is fully biologically active when compared to standard. The specific activity as determined in a viral resistance assay (human "Wish" cell line and VSV virus or the monkey VERO cell line with EMCV virus) was found to be 32 x 10⁶ IU/ mg.
Recombinant Human IFN-β 1b produced from E.Coli is a single, non-glycosylated human Interferon-β 1b polypeptide chain containing 165 amino acids and having a molecular mass of 18.51 kDa. The Interferon-β 1b gene was cloned from human fibroblasts. IFN-β 1b is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty