Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cortisol Human/Mouse/Rat Elisa Kit, BioVision

Supplier: BioVision
K7430-100
Sensitive, Colorimetric Assay

  • Detection method- Absorbance (450 nm)
  • Species reactivity- Human, mouse, rat
  • Application- Quantitative detection of Cortisol

BioVision’s Cortisol Enzyme-Linked Immunosorbent Assay (ELISA) Kit is an in vitro assay for the quantitative measurement of Cortisol in serum and plasma. Cortisol is a steroid hormone released from the adrenal cortex in response to the hormone ACTH. It increases blood pressure and blood sugar levels, and suppresses the immune system. Cortisol acts through specific intracellular receptors and has effects on numerous physiologic systems, including immune system, glucose-counter regulation, vascular tone, substrate utilization and bone metabolism. It exists in the blood as either a free form or bound to corticosteroid-binding globulin (CBG). The amount of Cortisol present in serum undergoes diurnal variation, with the highest levels present in the early morning and lower levels at night. BioVision’s Cortisol ELISA kit is a competitive enzyme immunoassay kit. The assay employs an antibody specific for cortisol coated on a 96-well plate. Cortisol in a sample or a HRP molecule which has Cortisol covalently attached to it is bound to the wells by immobilized antibody in a competitive manner. After a simultaneous incubation, the excess reagents are washed and a TMB substrate is added to the wells. Color develops in proportion to the amount of bound Cortisol HRP conjugate, and the intensity is inversely proportional to the concentration of Cortisol in the sample. Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. Sensitivity of the kit is 1 ng/ml and detection range is from 5 ng/ml to 480 ng/ml. The cross-reactivity of this ELISA assay is 6.8% for prednisolone, 4.22% for cortisone, and <0.1% for other hormones, including 11-deoxycortisol, estradiol, testosterone, and progesterone. The intra-assay reproducibility as measured by the coefficient of variation (CV) is < 8 % & inter-assay has CV < 12 %.




 

ORDER

Expand All / Collapse All
No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
100 Assays K7430-100 10013-298 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Storage Conditions Refer to data sheet
Species Reactivity Human, mouse, rat
Sample Type Serum and plasma; Cell culture supernatant

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty