Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Endothelin-1 Elisa Kit, BioVision

Supplier: BioVision
K7429-100
Sensitive, Colorimetric Assay

  • Detection method- Absorbance (450 nm)
  • Species reactivity- Human, mouse, rat, cow, dog, pig and rabbit
  • Application- Quantitative detection of Endothelin-1

BioVision’s Endothelin-1 (ET-1) ELISA kit is a competitive immunoassay for the quantitative determination of Endothelin-1 in biological fluids. ET-1 has identical amino acid sequence for human, mouse, rat, cow, dog, pig, and rabbit. The basal circulating level of ET-1 is reported to be < 1 to 3pg/mL but is known to be elevated in atrial and pulmonary hypertensions, atherosclerosis, congestive heart failure, cancer and variably related to lung diseases such as COPD and asthma. Samples and standards are added to wells coated with a monoclonal antibody specific for ET-1 and incubated. The plate is washed, leaving only bound ET-1 on the plate. A solution of HRP labeled monoclonal antibody to ET-1 is then added. This binds the ET-1 captured on the plate. The plate is then incubated. The plate is washed to remove excess HRP labeled antibody. TMB Substrate solution is added. The substrate generates a blue color when catalyzed by the HRP. Stop solution is added to stop the substrate reaction. The resulting yellow color is read at 450 nm. The amount of signal is directly proportional to the level of ET-1 in the sample. Sensitivity: 0.4 pg/ml (range: 0.78-100 pg/ml). The intra-Assay reproducibility is CV<10% & inter-Assay is CV<12%. Cross Reactivity: ET-1: 100%, ET-2: 21%, ET-3: 3.6%, human Big ET-1: <0.1%, rat Big ET-1: <0.1%, human Big ET-2: <0.1%, human Big ET-3 amide: <0.1%.




 

ORDER

Expand All / Collapse All
No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
100 Assays K7429-100 10013-786 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Storage Conditions 4°C
Species Reactivity Human, mouse, rat, cow, dog, pig and rabbit
Sample Type Serum, plasma, culture supernatants

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty