Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Progesterone Elisa Kit, BioVision

Supplier: BioVision

K7416-100
10013-784EA 736.45 USD InStock
10013-784
Progesterone Elisa Kit, BioVision
Assays ELISAs
Sensitive, Colorimetric Assay

  • Detection method- Absorbance (405 nm)
  • Species reactivity- all
  • Application- Quantitative detection of progesterone

BioVision’s Progesterone ELISA kit is a competitive immunoassay for the quantitative determination of Progesterone in biological fluids. The kit uses a polyclonal antibody to Progesterone to bind, in a competitive manner, Progesterone in a sample or Progesterone which has an alkaline phosphatase molecule covalently attached to it. After a simultaneous incubation at room temperature the excess reagents are washed away and substrate is added. After a short incubation time the enzyme reaction is stopped and the yellow color generated is read on a microplate reader at 405 nm. The intensity of the bound yellow color is inversely proportional to the concentration of Progesterone in either standards or samples. The measured optical density for the samples is used to calculate the concentration of Progesterone in the sample. Sensitivity: 8.57 pg/ml (range: 15.62-500 pg/ml). The intra-Assay reproducibility is CV<10% & inter-Assay is CV<12%. Cross Reactivity: Progesterone:100%, 5-Pregnane-3,20-dione: 100%, 17-OH-Progesterone: 3.46%, 5 alpha-Pregnen-3ß-o1-20-one: 1.43%, Corticosterone: 0.77%, 4-Androstene-3,17-dione: 0.28%, Deoxycorticosterone: 0.056%, DHEA: 0.013%, 17ß-Estradiol: <0.001%, Estrone: 0.001%, Estriol: <0.001%, Testosterone: <0.001%, Hydrocortisone: <0.001%, 5 alpha-Pregnane-3,20-diol: <0.001%, Danazol: <0.001%.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty