Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Resistin Human Serum Elisa Kit, BioVision

Supplier: BioVision

K4919-100
10013-742
Resistin Human Serum Elisa Kit, BioVision
  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects human Resistin in the range of 0.125–8 ng/ml resistin/ml. The lowest level of resistin that can be detected by this assay is 100 pg/ml.

Obesity is a well-known risk factor of type 2 diabetes mellitus and is strongly associated with insulin resistance. Resistin (also called FIZZ3/ADSF) is an adipocyte-derived peptide first identified during a search for targets of thiazolidinediones. Serum concentrations of resistin are markedly increased in obese mice and are decreased by treatment with thiazolidinediones. It was also found that administration of an anti-resistin antibody increases insulin-stimulated glucose uptake in obese mice and that treatment of normal mice with recombinant resistin impairs insulin action. Thus, resistin might link obesity with insulin resistance and diabetes in mice models. However, subsequent studies in rodent models have produced disparate findings on the role of resistin in obesity and insulin resistance. In humans, while the expression of resistin in human adipocytes is very low compared with that seen in rodents and does not differ between normal, insulin-resistant or type 2 diabetic individuals, a more recent study using a large size of case suggests that the plasma resistin levels are increased in type 2 diabetes. Therefore determination of the plasma resistin levels may be important for understanding onsets of metabolic diseases such as type 2 diabetes or obesity. This assay is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of human resistin in biological fluids. A monoclonal antibody specific for resistin has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, resistin is recognized by the addition of a biotinylated polyclonal antibody specific for resistin (Detection Antibody). After removal of excess biotinylated antibody, HRP labeled streptavidin (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of resistin in the samples. The assay range is 0.125–8 ng/ml resistin/ml. The lowest level of resistin that can be detected by this assay is 100 pg/ml.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty