Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vaspin Human Serum Elisa Kit, BioVision

Supplier: BioVision

K4917-100
10013-738EA 860.05 USD InStock
10013-738
Vaspin Human Serum Elisa Kit, BioVision
Assays ELISAs
  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects human Vaspin in the range of 0.016 – 1 ng/ml Vaspin/ml. The lowest level of vaspin that can be detected by this assay is 12 pg/ml.

Vaspin, designated as visceral adipose tissue-derived serpin, is a serpin whose expression is restricted to visceral adipose tissue. Vaspin seems to retain the serpin signature conformation fold consisting of three β-sheets and nine α-helices as well as a reactive loop site that interacts with its cognate serine protease unidentified hitherto. Vaspin is remarkably upregulated at high-fat high-sucrose (HFHS) diet and at multiple metabolic dysfunctions like obesity or insulin resistance. Administration of thiazolidinedione (TZD) caused a significant induction of serum vaspin in mice. These facts imply that upregulation of vaspin may be a compensatory response to antagonize the action of other unknown proteases upregulated in obesity or insulin resistance. Indeed, administration of recombinant vaspin to obese mice fed with HFHS chow improved glucose tolerance and insulin sensitivity. Vaspin seems to directly act on white adipose tissue. This assay is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of human vaspin in biological fluids. A monoclonal antibody specific for vaspin has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, vaspin is recognized by the addition of a purified polyclonal antibody specific for vaspin (Detection Antibody). After removal of excess polyclonal antibody, HRP conjugated anti-rabbit IgG (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of vaspin in the samples. The assay range is 0.016 – 1 ng/ml Vaspin/ml. The lowest level of vaspin that can be detected by this assay is 12 pg/ml.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty