Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Angptl6 Human Serum Elisa Kit, BioVision

Supplier: BioVision
K4916-100
  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects human ANGPTL6 in the range of 1.56 – 100 ng/ml ANGPTL6/ml. The lowest level of ANGPTL6 that can be detected by this assay is 1.2 ng/ml.

Seven angiopoietin-like proteins (ANGPTLs) share the characteristic protein structure of the angiopoietin family (ANG), but differ in their inability to bind antiopoietin receptor, Tie-2. ANGPTL6 was originally named angiopoietin-related growth factor (AGF) having an N-terminal coiled-coil-like domain and a C-terminal fibrinogen-like domain, both of which are conserved in ANG. It is a circulating protein secreted by liver that induces angiogenesis by direct effect of epidermal ANGPTL6 on endothelial cells and proliferation of skin cells, and thereby promotes wound healing. 80% of Angptl6 -/- mice died at about embryonic day 13. The surviving null mice developed marked obesity, lipid accumulation in skeletal muscle and liver, and insulin resistance accompanied by reduced energy expenditure relative to controls. Conversely, mice with constitutive overexpression of ANGPTL6 showed leanness and increased insulin sensitivity resulting from increased energy expenditure, and were also protected from high-fat diet-induced obesity, insulin resistance, and non-adipose tissue steatosis. This assay is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of human ANGPTL6 in biological fluids. A monoclonal antibody specific for ANGPTL6 has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, ANGPTL6 is recognized by the addition of a purified polyclonal antibody specific for ANGPTL6 (Detection Antibody). After removal of excess polyclonal antibody, HRP conjugated anti-rabbit IgG (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of ANGPTL6 in the samples. The assay range is 1.56 – 100 ng/ml ANGPTL6/ml. The lowest level of ANGPTL6 that can be detected by this assay is 1.2 ng/ml.




 

ORDER

Expand All / Collapse All
No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
100 Assays K4916-100 10013-736 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Storage Conditions +4°C
Species Reactivity Mammalian samples
Sample Type Cell and tissue culture supernatants, urine, plasma, serum, as well as many other biological fluids

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty