Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Angptl3 Mouse/Rat Serum Elisa Kit, BioVision

Supplier: BioVision
K4915-100
  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects mouse ANGPTL3 in the range of 0.016 – 1 ng/ml ANGPTL3/ml. The lowest level of ANGPTL3 that can be detected by this assay is 15 pg/ml.

The angiopoietins are a family of growth factors that are specific for vascular endothelium. The full-length cDNA encoding angiopoietin-like protein 3 (ANGPTL3) from a human fetal liver/spleen cDNA library has 460-amino acid and the characteristic structure of angiopoietins: a signal peptide, an extended helical domain predicted to form dimeric or trimeric coiled-coils, a short linker peptide, and a globular fibrinogen-like domain (FLD). Human ANGPTL3 shares 76% amino acid sequence identity with mouse Angptl3. Northern blot analysis of human tissues showed a preferential expression of 4 ANGPTL3 transcripts being 4.5, 3.0, 2.8, and 1.7 kb in liver. ANGPTL3 can induce angiogenesis in the rat corneal assay. The FLD alone was sufficient to induce endothelial cell adhesion and in vivo angiogenesis. Microarray analysis showed that mouse hematopoietic stem cell (HSC)-supportive fetal liver CD3-positive cells expressed Angptl2 and Angptl3. The ANGPTL3 ELISA Kit is to be used for the in vitro quantitative determination of mouse/ rat ANGPTL3 in biological fluids. This assay is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of mouse or rat ANGPTL3 in biological fluids. A polyclonal antibody specific for ANGPTL3 has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, ANGPTL3 is recognized by the addition of a biotinylated polyclonal antibody specific for ANGPTL3 (Detection Antibody). After removal of excess biotinylated antibody, HRP labeled streptavidin (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of ANGPTL3 in the samples. The assay range is 0.016 – 1 ng/ml ANGPTL3/ml. The lowest level of ANGPTL3 that can be detected by this assay is 15 pg/ml.




 

ORDER

Expand All / Collapse All
No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
100 Assays K4915-100 10013-734 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Storage Conditions +4°C
Species Reactivity Mammalian samples
Sample Type Cell and tissue culture supernatants, urine, plasma, serum, as well as many other biological fluids

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty