Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Angptl3 Human Serum Elisa Kit, BioVision

Supplier: BioVision

K4914-100
10013-732EA 939.74 USD InStock
10013-732
Angptl3 Human Serum Elisa Kit, BioVision
Assays ELISAs
  • Detection method- Absorbance (450 nm)
  • Sample type-
  • Species reactivity- Mammalian
  • Applications- This assay detects human ANGPTL3 in the range of 0.156–10 ng/ml ANGPTL3/ml. The lowest level of ANGPTL3 that can be detected by this assay is 75 pg/ml.

The angiopoietins are a family of growth factors that are specific for vascular endothelium. The full-length cDNA encoding angiopoietin-like protein 3 (ANGPTL3) from a human fetal liver/spleen cDNA library has 460-amino acid and the characteristic structure of angiopoietins: a signal peptide, an extended helical domain predicted to form dimeric or trimeric coiled-coils, a short linker peptide, and a globular fibrinogen-like domain (FLD). Human ANGPTL3 shares 76% amino acid sequence identity with mouse Angptl3. Northern blot analysis of human tissues showed a preferential expression of 4 ANGPTL3 transcripts being 4.5, 3.0, 2.8, and 1.7 kb in liver. ANGPTL3 can induce angiogenesis in the rat corneal assay. The FLD alone was sufficient to induce endothelial cell adhesion and in vivo angiogenesis. Microarray analysis showed that mouse hematopoietic stem cell (HSC)-supportive fetal liver CD3-positive cells expressed Angptl2 and Angptl3. The ANGPTL3 ELISA Kit is to be used for the in vitro quantitative determination of human ANGPTL3 in biological fluids. This assay is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of human ANGPTL3 in biological fluids. A monoclonal antibody specific for ANGPTL3 has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, ANGPTL3 is recognized by the addition of a purified polyclonal antibody specific for ANGPTL3 (Detection Antibody). After removal of excess polyclonal antibody, HRP conjugated anti-rabbit IgG (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of ANGPTL3 in the samples.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty