Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rbp4 Human Elisa Kit, BioVision

Supplier: BioVision

K4912-100
10013-728
Rbp4 Human Elisa Kit, BioVision
No cross-reactivity with mouse/rat RBP4

  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects human RBP4 in the range of 0.39 – 25 ng RBP4/ml and a detection limit of 380 pg/ml (based on adding two standard deviations to the mean value of the (50) zero standards).

Retinol binding protein (RBP) 4 is the only specific transport protein for vitamin A in the circulation whose function is to deliver vitamin to target tissues. In obesity and type 2 diabetes, expression of Glut4 is significantly impaired in adipocytes. Glucose transport via Glut4 is the rate-limiting step for glucose use by muscle and adipose tissue. Adipocyte-specific deletion of Gluts leads to notable elevation of RBP4 causing systemic insulin resistance, and that reduction of RBP4 improves insulin resistance. This identified a novel role of RBP4 in regulating insulin action and RBP4 is recorded as an adipocyte-derived hormone. The RBP4 (human) ELISA Kit is to be used for the in vitro quantitative determination of human RBP4 in biological fluids. This assay is a sandwich ELISA which utilizies a 96-well microtiter plate which was pre-coated with a monoclonal antibody and a purified polyclonal detection antibody. A HRP-conjugated anti-IgG, peroxidase and TMB (3,3’,5,5’-tetramethylbenzidine) is added to generate a color intensity directly proportional to the concentration of RBP4 in the samples. This ELISA is specific for the measurement of natural and recombinant human RBP4. It does not cross-react with mouse RBP4, rat RBP4, human adiponectin, human resistin, human vaspin, human clusterin, human leptin, human IL-33, human GPX3, human progranulin, human FABP4, human ANGPTL3, human ANGPTL4, human ANG1, human ANG2, human Nampt, mouse Nampt. The assay range is 0.39 – 25 ng RBP4/ml and a detection limit of 380 pg/ml (based on adding two standard deviations to the mean value of the (50) zero standards).

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty