Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rbp4 Human Competitive Elisa Kit, BioVision

Supplier: BioVision

K4911-100
10013-726EA 860.05 USD InStock
10013-726
Rbp4 Human Competitive Elisa Kit, BioVision
Assays ELISAs
No cross-reactivity with mouse/rat RBP4

  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mammalian
  • Applications- This assay detects human RBP4 in the range of 0.001 – 5 µg RBP4/ml and a detection limit of 1 ng/ml (based on adding two standard deviations to the mean value of the (50) zero standards).

Retinol binding protein (RBP) 4 is the only specific transport protein for vitamin A in the circulation whose function is to deliver vitamin to target tissues. In obesity and type 2 diabetes, the expression of Glut4 is significantly impaired in adipocytes. Glucose transport via Glut4 is the rate-limiting step for glucose use by muscle and adipose tissue. Adipocyte-specific deletion of Gluts leads to notable elevation of mouse RBP4 causing systemic insulin resistance, and that reduction of RBP4 improves insulin resistance. This identified a novel role of RBP4 in regulating insulin action and RBP4 is recorded as an adipocyte-derived hormone. The RBP4 (human) ELISA Kit is to be used for the in vitro quantitative determination of human RBP4 in serum, urine and cell culture supernatant. This assay is a competitive ELISA which utilizies a 96-well microtiter plate which was pre-coated with a human RBP4. A purified polyclonal which recognizing native human RBP4 reacts with a series of predetermined recombinant human RBP4 standard proteins or the test samples under competition in the human RBP4-coated plate. Their relative reactivity is plotted with that of the standard proteins. This ELISA is specific for the measurement of natural and recombinant human RBP4. It does not cross-react with mouse RBP4, rat RBP4, human adiponectin, rat adiponectin, human resistin, human vaspin, human clusterin, human leptin, human IL-23, human IL-33, human GPX3, human Nampt, human ANG1, human ANG2, human ANGPTL3, human ANGPTL4, human ANGPTL6, human FABP4, human RELM-β, rat RELM-α, mouse Nampt, human PAI-1. The assay range is 0.001 – 5 µg RBP4/ml and a detection limit of 1 ng/ml (based on adding two standard deviations to the mean value of the (50) zero standards).

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty