Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Progranulin Mouse Elisa Kit, BioVision

Supplier: BioVision

K4734-100
10013-714EA 751.9 USD InStock
10013-714
Progranulin Mouse Elisa Kit, BioVision
Assays ELISAs
  • Detection method- Absorbance (450 nm)
  • Species reactivity- Mouse

BioVision’s Progranulin (mouse) ELISA kit is a sandwich Enzyme Linked-Immunosorbent Assay (ELISA) for quantitative determination of human progranulin in biological fluids. A polyclonal antibody specific for progranulin has been precoated onto the 96-well microtiter plate. Standards and samples are pipetted into the wells for binding to the coated antibody. After extensive washing to remove unbound compounds, progranulin is recognized by the addition of a biotinylated polyclonal antibody specific for progranulin (Detection Antibody). After removal of excess biotinylated antibody, HRP labeled streptavidin (Detector) is added. Following a final washing, peroxidase activity is quantified using the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). The intensity of the color reaction is measured at 450 nm after acidification and is directly proportional to the concentration of progranulin in the samples.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty