Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cxcl12/Sdf-1 Alpha Human Elisa Kit, BioVision

Supplier: BioVision

K4387-100
10013-078EA 757.05 USD InStock
10013-078
Cxcl12/Sdf-1 Alpha Human Elisa Kit, BioVision
Assays ELISAs
Sensitive, Colorimetric Assay

  • Detection method- Absorbance (450 nm)
  • Species reactivity- human
  • Application- Quantitative protein detection, establishing normal range, validation of antibody array results, strip plates and additional reagents allow for use in multiple experiments.

BioVision’s Human SDF-1 (Stromal cell-derived factor-1) alpha ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human SDF-1 alpha. This assay employs an antibody specific for human SDF-1 alpha coated on a 96-well plate. Standards and samples are pipetted into the wells and SDF-1 alpha present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated anti-human SDF-1 alpha antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of SDF-1 alpha bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. This ELISA kit shows no cross-reactivity with any of the cytokines tested e.g., human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-γ, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1α, MIP-1 β, MIP-1δ, PARC, PDGF, RANTES, SCF, TARC, TGF-β, TIMP-1, TIMP-2, TNF-α, TNF-β, TPO, VEGF. Sensitivity: 80 pg/ml. Detection Range: 80 pg/ml – 6,000 pg/ml. The intra-Assay reproducibility is CV<10% & inter-Assay is CV<12%.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty