Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xanthine/Hypoxanthine Assay Kit, BioVision

Supplier: BioVision

K685-100
10013-774EA 324.45 USD InStock
10013-774
Xanthine/Hypoxanthine Assay Kit, BioVision
Assays Protein Assays
Sensitive Assay, HTS

  • Detection method- Absorbance (570 nm) or Fluoroescence (Ex/Em = 535/587 nm)
  • Application- The kit can detect Xanthine/Hypoxanthine levels as low as 0.4 µM in various biological samples.

Xanthine, a catabolic product of purine metabolism, is present in body fluids, muscle tissue and certain plants. Structurally like caffeine, Xanthine has a stimulant effect and is used clinically to treat the congestive diseases such as asthma and chronic obstructive pulmonary disease. Xanthine is metabolized into uric acid and superoxide by Xanthine oxidase. Xanthine oxidase deficiency causes the rare genetic disorder-Xanthinuria, and leads to Xanthine accumulation in urine and blood, which ultimately progresses to renal failure. Recent studies show that Xanthine levels are elevated following ischemic injury, thus Xanthine can serve as a useful marker for tissue hypoxia. Early detection of Xanthine alteration in biological fluids is crucial for metabolic studies and for diagnostic and therapeutic monitoring. In BioVision’s Xanthine/Hypoxanthine Assay kit, Xanthine/Hypoxanthine is specifically oxidized by the Xanthine Enzyme Mix to form an intermediate, which reacts with Developer & Probe to form a product that can be measured colorimetrically (λ = 570 nm) or fluorometrically (Ex/Em = 535/587 nm). Xanthine/Hypoxanthine Assay kit is rapid, simple and sensitive. This high-throughput suitable assay kit can detect Xanthine levels as low as 0.4 µM in various biological samples.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty