Free Glycerol Colorimetric Assay Kit II, BioVision

Supplier: BioVision
AB155899-100TEST
10013-764EA 851.41 USD
10013-764
Free Glycerol Colorimetric Assay Kit II, BioVision
Assays Protein Assays
For samples having reducing substances

  • Detection method- Absorbance (450 nm)
  • Application- Kit can detect less than 20 µM of Free Glycerol in various biological samples.

Glycerol is a central component of most lipids. It acts as a backbone for triglycerides and phospholipids, which play an important role in metabolism and cell membrane structure. Due to its low toxicity, glycerol is widely used in the pharmaceutical, food and cosmetic industries. Biovision’s Free Glycerol Assay kit II is suitable for measuring free glycerol levels in samples that contain reducing substances, which may interfere with oxidase-based assays. In this assay, Glycerol in the presence of Glycerol Enzyme Mix is converted to an intermediate, which reduces a colorless Probe to a colored product with strong absorbance at 450 nm. Free Glycerol Assay Kit II is simple, rapid and high-throughput adaptable. This assay kit can detect less than 20 mM of free Glycerol in various biological samples.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR