Would you like to log in again?
Cholesterol Detection Kit Cell-Based, BioVision
Supplier: BioVision
K587-100
10013-760EA 224 USD InStock
10013-760
Cholesterol Detection Kit Cell-Based, BioVision
Assays Protein Assays
Sensitive, simple staining for cellular unesterified cholesterol.
  • Detection method- Light and Fluorescence Microscopy (Ex/Em = 340-380/385-470 nm).
  • Applications- Screen/study/characterize stimulators/inhibitors that affect cholesterol transport and localization in a variety of cells.
Cholesterol is an essential structural component of animal cell membranes. It is required to maintain membrane structural integrity and fluidity. Cholesterol also serves as a precursor for the biosynthesis of several hormones, bile acid and vitamin D. Plasma membrane contains majority of the cellular cholesterol (80-90%) whereas little cholesterol resides in endoplasmic reticulum and in mitochondrial membrane. Transport of intracellular cholesterol within the cells to different compartments is through vesicular and non-vesicular pathways. Defects in these transport processes can alter cellular cholesterol metabolism resulting in pathological conditions. Filipin III is widely used as a probe for sterol localization in membranes. Interaction with cholesterol alters Filipin absorption and fluorescence spectra. This assay provides a simple, easy to perform, histochemical method of identification of unesterified cholesterol.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty