Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hdl Uptake Assay Kit Fluorometric, BioVision

Supplier: BioVision
K586-100
Only kit on the market to measure HDL uptake

  • Detection method- Fluorescence (Ex/Em 540/575 nm) Applications:
  • Measure HDL uptake in different cell types
  • Screen, study or characterize stimulators or inhibitors of HDL uptake

High-density lipoprotein (HDL) consists of a protein shell, containing apolipoproteins A-I and A-II, and a hydrophobic core consisting of cholesterol and triglycerides surrounded by phospholipids and cholesterol esters. HDL transports cholesterol to the liver or steroidogenic organs such as adrenals, ovary, and testes by both direct and indirect pathways. HDL-Cholesterol is removed by HDL receptors such as scavenger receptor BI (SR-BI), which mediate the selective uptake of cholesterol from HDL. Clinically, high levels of HDL has been associated with cardiovascular health. This is due to the ability of HDL particle to transport cholesterol from lipid-laden macrophages of atherosclerotic arteries to the liver for secretion into the bile by a process called as reverse cholesterol transport. BioVision’s HDL Uptake Assay Kit contains a fluorescently-labeled HDL that can be measured when taken up by cells. Unlabeled-HDL is included in the kit for assay validation.




 

ORDER

Expand All / Collapse All
No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
100 Assays K586-100 10013-758 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Storage Conditions -20°C
Species Reactivity Mammalian samples
Sample Type Cells

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty