Would you like to log in again?
Ready-To-Use Ihc Kit Biotin Free, BioVision
Supplier: BioVision
K405-50
10013-702
Ready-To-Use Ihc Kit Biotin Free, BioVision
Highly Sensitive, IHC/ICC Kit
  • Species reactivity- Mouse, rat, and rabbit
  • Applications- A biotin/avidin free system that stains membrane, cytoplasmic and nuclear antigens. Immunohistochemistry (IHC) and immunocytochemistry (ICC).
Immunohistochemistry (IHC)/Immunocytochemistry (ICC) is the localization of antigens in tissue sections/cells by the use of labeled antibodies as specific reagents through antigen-antibody interactions that are visualized by a marker such as fluorescent dye, enzyme, radioactive element or colloidal gold. Several IHC techniques are commonly used such as labeled biotin secondary antibody streptavidin-peroxidase, HRP anti-HRP, ABC catalyzed signal amplification, polymer system (one or two steps) etc. to detect antigens in tissues and cells. BioVision’s Ready-to-use IHC kit employs polymer technology that provides increased sensitivity and detection. IBSC One-Step anti-mouse, Rat and Rabbit IgG (H+L) is a biotin/avidin free system that stains membrane, cytoplasmic and nuclear antigens. It provides the user with a rapid and easy to use IHC detection system.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty