Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hdac8 Inhibitor Screening Kit, BioVision

Supplier: BioVision

K368-100
10013-692
Hdac8 Inhibitor Screening Kit, BioVision
Simple Assay, HTS

  • Detection method- Fluorescence (Em/Ex = 380/500 nm)
  • Applications- Suitable for high throughput screening of HDAC-8 inhibitors

Histone deacetylases (HDACs) represent a large family of enzymes identified as key regulators of nucleosomal histone acetylation, a major event that controls eukaryotic gene transcription and are classified into three groups. They are believed to be involved in important biological activities, such as cell differentiation, proliferation, apoptosis, and senescence. In BioVision’s HDAC-8 Inhibitor screening Kit, HDAC-8 deacetylates the substrate (R-H-K(Ac)-K(Ac)-AFC) followed by an enzymatic cleavage to release the quenched fluorescent group (AFC), which can be detected at Em/Ex = 380/500 nm. In the presence of an HDAC-8 inhibitor, the deacetylation will be impeded which prevents the cleavage of the substrate to release AFC. The kit provides a rapid, simple, sensitive, and reliable test, which is also suitable for high throughput screening of HDAC-8 inhibitors. Trichostatin A is included as a control inhibitor to compare the efficacy of the test inhibitors.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty