Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ffactor Ixa Activ Assay Kit Fluorometric, BioVision

Supplier: BioVision

K364-100
10013-690
Ffactor Ixa Activ Assay Kit Fluorometric, BioVision
Simple, Highly Sensitive Assay

  • Detection method- Fluorescence (Ex/Em = 360/450 nm
  • Applications:
  • - Determine activity of purified Factor IXa
  • - Detect the activity of Factor IXa in plasma and serum samples

The coagulation Factor IX (or Christmas factor) (EC 3.4.21.22) is a vitamin K-dependent serine protease. Factor IX is produced as an inactive precursor and is activated via cleavage by either factor XIa (contact pathway) or factor VIIa (tissue factor pathway). In the presence of calcium ions and negatively charged membrane phospholipids, activated factor IX (FIXa) then binds to the activated Factor VIII (FVIIIa) and proteolytically activates factor X (FX) to factor Xa (FXa). BioVision’s Factor IXa Activity Assay kit is based on the ability of FIXa to generate FXa. The generated FXa proteolytically cleaves a synthetic substrate and releases a fluorophore, AMC, which can be easily quantified by fluorescence microplate reader. The assay is simple, rapid and can detect activity as low as 10 pg of FIXa in a variety of samples.

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty