Would you like to log in again?
PCNA, human recombinant, BioVision
Supplier: BioVision
4835-5
10007-184EA 365.65 USD InStock
10007-184
PCNA, human recombinant, BioVision
Proteins and Peptides
>90% Pure Recombinant human PCNA
Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase δ auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-δ to play a fundamental role in the initiation of cell proliferation. Recombinant PCNA was expressed with a 6x His tag in sf9 insect cells and purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty