Would you like to log in again?
Portable Balance, CL Series, Ohaus®
Supplier: OHAUS SCALE CORP
80010610 80010611 80010612
Reliable, easy-to-use balance for basic weighing.
  • The integrated load cell lock protects the weighing cell from damage during transportation and while in storage
  • The CL is battery operated and automatically shuts off after a certain period of non-use to preserve battery life
  • The low profile CL has a stackable design which allows the balance to be stored securely and orderly
The CL Series is a lightweight, portable balance for industrial, jewelry and home use applications. This economical balance with a sleek compact design and reliable performance has all the tools to meet your basic weighing requirements.
ORDER
Expand All / Collapse All
Model Weighing Capacity Pan/Platform Size Readability Supplier No. VWR Catalog Number Unit Price Quantity
CL201 200 g 120 mm (4.7") 0.1 g 80010610 89425-812 Each Retrieving
CL2000 2000 g 120 mm (4.7") 1 g 80010611 89425-810 Each Retrieving
CL5000 5000 g 120 mm (4.7") 1 g 80010612 89425-814 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty