Would you like to log in again?
Collagen-III, human recombinant, BioVision
Supplier: BioVision
4797-5
10007-162
Collagen-III, human recombinant, BioVision
>95% Pure Recombinant human Collagen-III.
Collagen, a major component of the extraCellular matrix, is a fibrous protein that provides strength to tissues giving them structural integrity. It has been part of the medical, pharmaceutical and consumer products for over 100 years.DNA sequences encoding the human proα 1 (III) and both α and β subunits of the prolyl hydoxylase were co-expressed in the yeast Pichia Pastoris. Procollagen III was converted to mature collagen by controlled proteinase digestion.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty